Assay Details
| Gene Symbol: bmpr1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004585.1 | NP_001004585.1 | 9 - 10 | 1328 | 84 | |
| GenBank mRNA | BC081626.1 | - | 9 - 10 | 1328 | 84 | |
More Information
Set Membership: |
|

bmpr1ab
Genomic Map