Assay Details
| Gene Symbol: gcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005603.1 | NP_001005603.1 | 3 - 4 | 782 | 133 | |
| XM_005162569.4 | XP_005162626.1 | 4 - 5 | 788 | 133 | ||
| GenBank mRNA | AB099697.1 | - | 3 - 4 | 782 | 133 | |
| AB175671.1 | - | 4 - 5 | 751 | 133 | ||
| AY217729.1 | - | 3 - 4 | 820 | 133 | ||
| AY706754.1 | - | 3 - 4 | 474 | 133 | ||
| BC162304.1 | - | 3 - 4 | 780 | 133 | ||
More Information
Set Membership: |
|

gcm2
Genomic Map