Assay Details
| Gene Symbol: kctd15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006012.1 | NP_001006012.1 | 4 - 5 | 792 | 73 | |
| XM_005163051.4 | XP_005163108.1 | 4 - 5 | 1103 | 73 | ||
| XM_005163052.2 | XP_005163109.1 | 4 - 5 | 894 | 73 | ||
| GenBank mRNA | BC083478.1 | - | 4 - 5 | 792 | 73 | |
More Information
Set Membership: |
|

kctd15a
Genomic Map