Assay Details
| Gene Symbol: acot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006072.1 | NP_001006072.1 | 3 - 4 | 512 | 64 | |
| GenBank mRNA | BC083249.1 | - | 3 - 4 | 512 | 64 | |
| BC164121.1 | - | 3 - 4 | 537 | 64 | ||
More Information
Set Membership: |
|

acot8
Genomic Map