Assay Details
| Gene Symbol: acot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006072.1 | NP_001006072.1 | 5 - 6 | 855 | 61 | |
| GenBank mRNA | BC083249.1 | - | 5 - 6 | 855 | 61 | |
| BC164121.1 | - | 5 - 6 | 880 | 61 | ||
More Information
Set Membership: |
|

acot8
Genomic Map