Assay Details
| Gene Symbol: mvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007349.1 | NP_001007350.1 | 1 - 2 | 290 | 63 | |
| GenBank mRNA | BC085528.1 | - | 1 - 2 | 290 | 63 | |
| BC164491.1 | - | 1 - 2 | 118 | 63 | ||
More Information
Set Membership: |
|

mvk
Genomic Map