Assay Details
| Gene Symbol: thg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007455.1 | NP_001007456.1 | 4 - 5 | 905 | 148 | |
| XM_017359009.2 | XP_017214498.1 | 3 - 4 | 222 | 148 | ||
| GenBank mRNA | BC085384.1 | - | 4 - 5 | 905 | 148 | |
| BC164839.1 | - | 4 - 5 | 571 | 148 | ||
More Information
Set Membership: |
|

thg1l
Genomic Map