Assay Details
| Gene Symbol: gna11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007773.1 | NP_001007774.1 | 3 - 4 | 979 | 68 | |
| GenBank mRNA | BC085433.1 | - | 3 - 4 | 979 | 68 | |
| BC164178.1 | - | 3 - 4 | 506 | 68 | ||
More Information
Set Membership: |
|

gna11b
Genomic Map