Assay Details
| Gene Symbol: arhgef7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008624.1 | NP_001008624.1 | 15 - 16 | 1828 | 87 | |
| XM_017354714.2 | XP_017210203.1 | 15 - 16 | 2007 | 87 | ||
| XM_017354716.2 | XP_017210205.1 | 15 - 16 | 1640 | 87 | ||
| XM_017354718.2 | XP_017210207.1 | 15 - 16 | 1603 | 87 | ||
| XM_021474150.1 | XP_021329825.1 | 15 - 16 | 1713 | 87 | ||
| GenBank mRNA | BC086736.1 | - | 15 - 16 | 1828 | 87 | |
| BC165668.1 | - | 15 - 16 | 1747 | 87 | ||
| DQ656108.1 | - | 15 - 16 | 1614 | 87 | ||
| DQ656109.1 | - | 15 - 16 | 1782 | 87 | ||
More Information
Set Membership: |
|

arhgef7b
Genomic Map