Assay Details
Gene Symbol: ccdc191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009986.1 | NP_001009986.1 | 4 - 5 | 550 | 130 | |
XM_009297387.3 | XP_009295662.1 | 4 - 5 | 605 | 130 | ||
XM_017353826.2 | XP_017209315.1 | 4 - 5 | 448 | 130 | ||
GenBank mRNA | BC088387.1 | - | 4 - 5 | 550 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
