Assay Details
| Gene Symbol: bnip3la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012242.1 | NP_001012242.1 | 4 - 5 | 504 | 79 | |
| GenBank mRNA | AY424951.1 | - | 4 - 5 | 504 | 79 | |
| BC163834.1 | - | 4 - 5 | 516 | 79 | ||
| BC163857.1 | - | 4 - 5 | 516 | 79 | ||
More Information
Set Membership: |
|

bnip3la
Genomic Map