Assay Details
| Gene Symbol: trh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012365.2 | NP_001012365.2 | 2 - 3 | 240 | 62 | |
| GenBank mRNA | AY903312.1 | - | 2 - 3 | 62 | ||
| BC095193.1 | - | 2 - 3 | 240 | 62 | ||
More Information
Set Membership: |
|

trh
Genomic Map