Assay Details
| Gene Symbol: upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013301.1 | NP_001013319.1 | 4 - 5 | 454 | 118 | |
| XM_005157923.4 | XP_005157980.1 | 4 - 5 | 460 | 118 | ||
| XM_017351344.2 | XP_017206833.1 | 4 - 5 | 494 | 118 | ||
| GenBank mRNA | BC090681.1 | - | 4 - 5 | 454 | 118 | |
More Information
Set Membership: |
|

upp1
Genomic Map