Assay Details
| Gene Symbol: ywhae2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013341.2 | NP_001013359.1 | 1 - 2 | 209 | 82 | |
| XM_021477523.1 | XP_021333198.1 | 1 - 2 | 273 | 82 | ||
| GenBank mRNA | BC090759.1 | - | 1 - 2 | 213 | 82 | |
| BC165124.1 | - | 1 - 2 | 98 | 82 | ||
More Information
Set Membership: |
|

ywhae2
Genomic Map