Assay Details
| Gene Symbol: ywhae2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013341.2 | NP_001013359.1 | 4 - 5 | 725 | 81 | |
| XM_021477523.1 | XP_021333198.1 | 4 - 5 | 789 | 81 | ||
| GenBank mRNA | BC090759.1 | - | 4 - 5 | 729 | 81 | |
| BC165124.1 | - | 4 - 5 | 614 | 81 | ||
More Information
Set Membership: |
|

ywhae2
Genomic Map