Assay Details
| Gene Symbol: lifra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014306.2 | NP_001014328.2 | 13 - 14 | 1975 | 67 | |
| GenBank mRNA | BC091862.1 | - | 13 - 14 | 1979 | 67 | |
| BC165177.1 | - | 12 - 13 | 1805 | 67 | ||
More Information
Set Membership: |
|

lifra
Genomic Map