Assay Details
| Gene Symbol: lifra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014306.2 | NP_001014328.2 | 5 - 6 | 860 | 114 | |
| GenBank mRNA | BC091862.1 | - | 5 - 6 | 864 | 114 | |
| BC165177.1 | - | 4 - 5 | 690 | 114 | ||
More Information
Set Membership: |
|

lifra
Genomic Map