Assay Details
| Gene Symbol: lifra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014306.2 | NP_001014328.2 | 6 - 7 | 988 | 108 | |
| GenBank mRNA | BC091862.1 | - | 6 - 7 | 992 | 108 | |
| BC165177.1 | - | 5 - 6 | 818 | 108 | ||
More Information
Set Membership: |
|

lifra
Genomic Map