Assay Details
| Gene Symbol: angptl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014820.1 | NP_001014820.1 | 2 - 3 | 983 | 73 | |
| XM_005160416.4 | XP_005160473.1 | 3 - 4 | 1450 | 73 | ||
| GenBank mRNA | AB199647.1 | - | 2 - 3 | 983 | 73 | |
| BC163080.1 | - | 2 - 3 | 1025 | 73 | ||
| BC163101.1 | - | 2 - 3 | 1025 | 73 | ||
More Information
Set Membership: |
|

angptl1a
Genomic Map