Assay Details
| Gene Symbol: acer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017603.1 | NP_001017603.1 | 3 - 4 | 387 | 80 | |
| XM_005166840.4 | XP_005166897.1 | 3 - 4 | 509 | 80 | ||
| GenBank mRNA | BC092849.1 | - | 3 - 4 | 387 | 80 | |
| BC164606.1 | - | 2 - 3 | 241 | 80 | ||
More Information
Set Membership: |
|

acer1
Genomic Map