Assay Details
| Gene Symbol: ppa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017833.1 | NP_001017833.1 | 4 - 5 | 346 | 143 | |
| GenBank mRNA | BC092782.1 | - | 4 - 5 | 346 | 143 | |
| BC164460.1 | - | 4 - 5 | 330 | 143 | ||
More Information
Set Membership: |
|

ppa1b
Genomic Map