Assay Details
| Gene Symbol: ppa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017833.1 | NP_001017833.1 | 5 - 6 | 436 | 128 | |
| GenBank mRNA | BC092782.1 | - | 5 - 6 | 436 | 128 | |
| BC164460.1 | - | 5 - 6 | 420 | 128 | ||
More Information
Set Membership: |
|

ppa1b
Genomic Map