Assay Details
| Gene Symbol: rgra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017877.1 | NP_001017877.1 | 3 - 4 | 535 | 77 | |
| GenBank mRNA | BC092971.1 | - | 3 - 4 | 535 | 77 | |
| BC165377.1 | - | 3 - 4 | 397 | 77 | ||
| KT008426.1 | - | 3 - 4 | 363 | 77 | ||
More Information
Set Membership: |
|

rgra
Genomic Map