Assay Details
| Gene Symbol: ptbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020477.1 | NP_001018313.1 | 2 - 3 | 71 | 86 | |
| GenBank mRNA | BC095372.1 | - | 2 - 3 | 71 | 86 | |
More Information
Set Membership: |
|

ptbp1a
Genomic Map