Assay Details
| Gene Symbol: ptbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020477.1 | NP_001018313.1 | 3 - 4 | 108 | 122 | |
| XM_021478816.1 | XP_021334491.1 | 3 - 4 | 175 | 119 | ||
| GenBank mRNA | BC095372.1 | - | 3 - 4 | 108 | 122 | |
More Information
Set Membership: |
|

ptbp1a
Genomic Map