Assay Details
| Gene Symbol: ezra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020490.1 | NP_001018326.1 | 2 - 3 | 184 | 73 | |
| GenBank mRNA | BC054026.1 | - | 2 - 3 | 191 | 73 | |
| BC095359.1 | - | 2 - 3 | 184 | 73 | ||
| BC153484.1 | - | 2 - 3 | 189 | 73 | ||
| BC165851.1 | - | 1 - 2 | 132 | 73 | ||
More Information
Set Membership: |
|

ezra
Genomic Map