Assay Details
| Gene Symbol: ezra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020490.1 | NP_001018326.1 | 8 - 9 | 1039 | 149 | |
| GenBank mRNA | BC095359.1 | - | 8 - 9 | 1039 | 149 | |
| BC153484.1 | - | 8 - 9 | 149 | |||
| BC165851.1 | - | 7 - 8 | 987 | 149 | ||
More Information
Set Membership: |
|

ezra
Genomic Map