Assay Details
| Gene Symbol: ezra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020490.1 | NP_001018326.1 | 11 - 12 | 1426 | 72 | |
| GenBank mRNA | BC095359.1 | - | 11 - 12 | 1426 | 72 | |
| BC165851.1 | - | 10 - 11 | 1374 | 72 | ||
More Information
Set Membership: |
|

ezra
Genomic Map