Assay Details
| Gene Symbol: rhof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020642.1 | NP_001018478.1 | 2 - 3 | 399 | 64 | |
| GenBank mRNA | BC095646.1 | - | 2 - 3 | 399 | 64 | |
| BC165017.1 | - | 2 - 3 | 251 | 64 | ||
More Information
Set Membership: |
|

rhof
Genomic Map