Assay Details
| Gene Symbol: stx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024425.1 | NP_001019596.1 | 1 - 2 | 191 | 62 | |
| XM_009307637.3 | XP_009305912.1 | 1 - 2 | 208 | 62 | ||
| GenBank mRNA | BC095763.1 | - | 1 - 2 | 191 | 62 | |
More Information
Set Membership: |
|

stx18
Genomic Map