Assay Details
| Gene Symbol: sec62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025530.1 | NP_001020701.1 | 1 - 2 | 91 | 59 | |
| GenBank mRNA | BC097135.1 | - | 1 - 2 | 91 | 59 | |
More Information
Set Membership: |
|

sec62
Genomic Map