Assay Details
| Gene Symbol: u2af2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029956.1 | NP_001025127.1 | 1 - 2 | 455 | 76 | |
| XM_005157895.4 | XP_005157952.1 | 1 - 2 | 514 | 76 | ||
| GenBank mRNA | BC098548.1 | - | 1 - 2 | 455 | 76 | |
More Information
Set Membership: |
|

u2af2a
Genomic Map