Assay Details
| Gene Symbol: u2af2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029956.1 | NP_001025127.1 | 3 - 4 | 720 | 115 | |
| XM_005157895.4 | XP_005157952.1 | 3 - 4 | 779 | 115 | ||
| XM_021466696.1 | XP_021322371.1 | 3 - 4 | 801 | 115 | ||
| GenBank mRNA | BC098548.1 | - | 3 - 4 | 720 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021466696.1 transcripts.
Set Membership: |
|

u2af2a
Genomic Map