Assay Details
| Gene Symbol: u2af2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029956.1 | NP_001025127.1 | 9 - 10 | 1418 | 117 | |
| XM_005157895.4 | XP_005157952.1 | 9 - 10 | 1492 | 117 | ||
| XM_021466696.1 | XP_021322371.1 | 9 - 10 | 1514 | 117 | ||
| GenBank mRNA | BC098548.1 | - | 9 - 10 | 1418 | 117 | |
More Information
Set Membership: |
|

u2af2a
Genomic Map