Assay Details
| Gene Symbol: etsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037375.1 | NP_001032452.1 | 2 - 3 | 138 | 83 | |
| GenBank mRNA | BC124743.1 | - | 2 - 3 | 147 | 83 | |
| BC164765.1 | - | 1 - 2 | 108 | 83 | ||
| DQ021472.1 | - | 2 - 3 | 138 | 83 | ||
More Information
Set Membership: |
|

etsrp
Genomic Map