Assay Details
| Gene Symbol: etsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037375.1 | NP_001032452.1 | 5 - 6 | 531 | 89 | |
| XM_005158213.4 | XP_005158270.1 | 5 - 6 | 763 | 89 | ||
| GenBank mRNA | BC124743.1 | - | 5 - 6 | 540 | 89 | |
| BC164765.1 | - | 4 - 5 | 501 | 89 | ||
| DQ021472.1 | - | 5 - 6 | 531 | 89 | ||
More Information
Set Membership: |
|

etsrp
Genomic Map