Assay Details
| Gene Symbol: etsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037375.1 | NP_001032452.1 | 7 - 8 | 895 | 144 | |
| XM_005158213.4 | XP_005158270.1 | 7 - 8 | 1127 | 144 | ||
| GenBank mRNA | BC124743.1 | - | 7 - 8 | 904 | 144 | |
| BC164765.1 | - | 6 - 7 | 865 | 144 | ||
| DQ021472.1 | - | 7 - 8 | 895 | 144 | ||
More Information
Set Membership: |
|

etsrp
Genomic Map