Assay Details
| Gene Symbol: trmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037692.2 | NP_001032781.2 | 3 - 4 | 451 | 80 | |
| NM_001327757.1 | NP_001314686.1 | 4 - 5 | 713 | 80 | ||
| GenBank mRNA | BC107495.1 | - | 3 - 4 | 453 | 80 | |
| EH543034.1 | - | 4 - 5 | 666 | 80 | ||
| EH549833.1 | - | 4 - 5 | 735 | 80 | ||
More Information
Set Membership: |
|

trmo
Genomic Map