Assay Details
| Gene Symbol: prlhr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039526.1 | NP_001034615.1 | 1 - 2 | 476 | 93 | |
| XM_005167225.3 | XP_005167282.1 | 2 - 3 | 671 | 93 | ||
| GenBank mRNA | BC162438.1 | - | 1 - 2 | 521 | 93 | |
| BC162771.1 | - | 1 - 2 | 521 | 93 | ||
| DQ083991.1 | - | 1 - 2 | 476 | 93 | ||
| EU165204.1 | - | 1 - 2 | 482 | 93 | ||
More Information
Set Membership: |
|

prlhr2a
Genomic Map