Assay Details
| Gene Symbol: rnd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044396.2 | NP_001037861.1 | 3 - 4 | 461 | 84 | |
| GenBank mRNA | AY865571.1 | - | 3 - 4 | 319 | 84 | |
| BC076165.1 | - | 4 - 5 | 518 | 84 | ||
| BC122138.1 | - | 3 - 4 | 461 | 84 | ||
| BC165155.1 | - | 3 - 4 | 353 | 84 | ||
More Information
Set Membership: |
|

rnd1b
Genomic Map