Assay Details
| Gene Symbol: cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044883.1 | NP_001038348.1 | 10 - 11 | 1392 | 62 | |
| GenBank mRNA | BC171654.1 | - | 10 - 11 | 1486 | 62 | |
More Information
Set Membership: |
|

cftr
Genomic Map