Assay Details
| Gene Symbol: myt1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044899.2 | NP_001038364.1 | 9 - 10 | 2171 | 59 | |
| XM_005160722.4 | XP_005160779.1 | 11 - 12 | 2542 | 59 | ||
| XM_005160723.4 | XP_005160780.1 | 11 - 12 | 2542 | 59 | ||
| XM_009294757.3 | XP_009293032.1 | 11 - 12 | 2542 | 59 | ||
| XM_021468452.1 | XP_021324127.1 | 3 - 4 | 424 | 59 | ||
| GenBank mRNA | BC171626.1 | - | 10 - 11 | 2240 | 59 | |
More Information
Set Membership: |
|

myt1la
Genomic Map