Assay Details
| Gene Symbol: myt1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044899.2 | NP_001038364.1 | 4 - 5 | 604 | 158 | |
| XM_005160722.4 | XP_005160779.1 | 6 - 7 | 975 | 158 | ||
| XM_005160723.4 | XP_005160780.1 | 6 - 7 | 975 | 158 | ||
| XM_009294757.3 | XP_009293032.1 | 6 - 7 | 975 | 158 | ||
| GenBank mRNA | BC124350.1 | - | 6 - 7 | 934 | 158 | |
| BC171626.1 | - | 5 - 6 | 673 | 158 | ||
More Information
Set Membership: |
|

myt1la
Genomic Map