Assay Details
| Gene Symbol: rabl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044963.2 | NP_001038428.1 | 3 - 4 | 682 | 79 | |
| XM_009293419.3 | XP_009291694.1 | 2 - 3 | 731 | 79 | ||
| GenBank mRNA | BC134982.1 | - | 3 - 4 | 682 | 79 | |
More Information
Set Membership: |
|

rabl2
Genomic Map