Assay Details
| Gene Symbol: prmt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045042.1 | NP_001038507.1 | 2 - 3 | 238 | 86 | |
| XM_005164654.4 | XP_005164711.1 | 3 - 4 | 1068 | 86 | ||
| XM_005164655.4 | XP_005164712.1 | 3 - 4 | 1064 | 86 | ||
| XM_021475051.1 | XP_021330726.1 | 3 - 4 | 1061 | 86 | ||
| GenBank mRNA | BC162920.1 | - | 2 - 3 | 276 | 86 | |
| BC162940.1 | - | 2 - 3 | 276 | 86 | ||
More Information
Set Membership: |
|

prmt8b
Genomic Map