Assay Details
| Gene Symbol: mthfsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045099.1 | NP_001038564.1 | 7 - 8 | 790 | 105 | |
| GenBank mRNA | BC122258.1 | - | 7 - 8 | 790 | 105 | |
| BC171684.1 | - | 7 - 8 | 707 | 105 | ||
| BC171688.1 | - | 7 - 8 | 706 | 105 | ||
More Information
Set Membership: |
|

mthfsd
Genomic Map