Assay Details
| Gene Symbol: ripor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045108.2 | NP_001038573.1 | 2 - 3 | 243 | 75 | |
| XM_005159126.4 | XP_005159183.1 | 3 - 4 | 401 | 75 | ||
| XM_009293578.3 | XP_009291853.1 | 3 - 4 | 442 | 75 | ||
| XM_009293579.3 | XP_009291854.1 | 3 - 4 | 544 | 75 | ||
| XM_009293580.3 | XP_009291855.1 | 3 - 4 | 454 | 75 | ||
| XM_021467681.1 | XP_021323356.1 | 3 - 4 | 492 | 75 | ||
| XM_021467682.1 | XP_021323357.1 | 3 - 4 | 490 | 75 | ||
More Information
Set Membership: |
|

ripor1
Genomic Map