Assay Details
| Gene Symbol: slc6a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045287.2 | NP_001038752.2 | 9 - 10 | 1332 | 84 | |
| GenBank mRNA | BC116591.1 | - | 9 - 10 | 1324 | 84 | |
More Information
Set Membership: |
|

slc6a1a
Genomic Map