Assay Details
| Gene Symbol: gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045385.2 | NP_001038850.2 | 4 - 5 | 568 | 78 | |
| GenBank mRNA | BC122359.1 | - | 4 - 5 | 568 | 78 | |
More Information
Set Membership: |
|

gck
Genomic Map