Assay Details
| Gene Symbol: abi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076586.2 | NP_001070054.2 | 3 - 4 | 463 | 74 | |
| XM_005167480.4 | XP_005167537.1 | 3 - 4 | 490 | 74 | ||
| XM_005167483.4 | XP_005167540.1 | 3 - 4 | 490 | 74 | ||
| XM_009304513.3 | XP_009302788.1 | 3 - 4 | 490 | 74 | ||
| XM_021478763.1 | XP_021334438.1 | 3 - 4 | 490 | 74 | ||
| XM_021478764.1 | XP_021334439.1 | 3 - 4 | 493 | 74 | ||
| GenBank mRNA | BC122368.1 | - | 3 - 4 | 499 | 74 | |
| BC165556.1 | - | 3 - 4 | 497 | 74 | ||
More Information
Set Membership: |
|

abi2a
Genomic Map